4-Brom-N-butylbenzamid
APOSOR11942-1GEA
0
EUR
APOSOR11942-1G
APOSOR11942-5G
4-Brom-N-butylbenzamid
4-Brom-N-butylbenzamid
Formel:
C₁₁H₁₄BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00456169 CAS-Nummer: 78080-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...