N-(1-Naphthyl)-4-boronobenzolsulfonamid

Lieferant: Apollo Scientific

Synonyme: (4-(N-(Naphthalen-1-yl)sulphamoyl)phenyl)boronic acid,4-(Naphth-1-ylsulphamoyl)benzeneboronic acid

OR11939-1G
APOSOR11939-1GEA 0 EUR
APOSOR11939-1G
N-(1-Naphthyl)-4-boronobenzolsulfonamid
N-(1-Naphthyl)-4-boronobenzolsulfonamid
Formel: C₁₆H₁₄BNO₄S
Molecular Weight: 327,1600 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Refrigerator
MDL: MFCD09800885
CAS-Nummer: 957120-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR