2-Brom-N-butylbenzolsulfonamid
APOSOR11697-1GEA
0
EUR
APOSOR11697-1G
APOSOR11697-5G
2-Brom-N-butylbenzolsulfonamid
2-Brom-N-butylbenzolsulfonamid
Formel:
C₁₀H₁₄BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800950 CAS-Nummer: 951885-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...