Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat

APOSOR11685-1GEA 0 EUR
APOSOR11685-1G
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Formel: C₁₂H₁₀N₂O₅
Molecular Weight: 262,2200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179429
CAS-Nummer: 72030-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR