8-Brom-6-nitroimidazo[1,2-a]pyridin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR11664-1GEA
17.6
EUR
APOSOR11664-1G
8-Brom-6-nitroimidazo[1,2-a]pyridin Hydrochlorid
8-Brom-6-nitroimidazo[1,2-a]pyridin Hydrochlorid
Formel:
C₇H₅BrClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800973 CAS-Nummer: 957120-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...