3-Chlor-5-(ethylcarbamoyl)phenylboronsäure
APOSOR11660-1GEA
0
EUR
APOSOR11660-1G
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BClNO₃ Molecular Weight: 227,4500 g/mol Schmelzpunkt: 107…110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09800857 CAS-Nummer: 957120-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...