3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific

Achtung

APOSOR11654-5GEA 832 EUR
APOSOR11654-5G APOSOR11654-1G
3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure
3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BClNO₃
Molecular Weight: 227,4500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800861
CAS-Nummer: 957120-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR