2-Brom-N-cyclohexylbenzolsulfonamid

Lieferant: Apollo Scientific

APOSOR11653-5GEA 473 EUR
APOSOR11653-5G APOSOR11653-1G
2-Brom-N-cyclohexylbenzolsulfonamid
2-Brom-N-cyclohexylbenzolsulfonamid
Formel: C₁₂H₁₆BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09800965
CAS-Nummer: 951883-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR