2-Brom-N-hexylbenzolsulfonamid

APOSOR11646-1GEA 0 EUR
APOSOR11646-1G APOSOR11646-5G
2-Brom-N-hexylbenzolsulfonamid
2-Brom-N-hexylbenzolsulfonamid
Formel: C₁₂H₁₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09800964
CAS-Nummer: 951883-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR