3-Chlor-6-(cyclohexylamino)pyridazin
Lieferant: Apollo Scientific
APOSOR11638-5GEA
350
EUR
APOSOR11638-5G
APOSOR11638-1G
3-Chlor-6-(cyclohexylamino)pyridazin
3-Chloro-6-(cyclohexylamino)pyridazine
Formel:
C₁₀H₁₄ClN₃ Schmelzpunkt: 160…165 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00968568 CAS-Nummer: 1014-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...