2-Brom-N-pentylbenzolsulfonamid

Lieferant: Apollo Scientific

APOSOR11633-1GEA 132 EUR
APOSOR11633-1G APOSOR11633-5G
2-Brom-N-pentylbenzolsulfonamid
2-Brom-N-pentylbenzolsulfonamid
Formel: C₁₁H₁₆BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09800967
CAS-Nummer: 951883-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR