3-Chlor-6-(3-methoxypropylamino)pyridazin

APOSOR11607-1GEA 0 EUR
APOSOR11607-1G APOSOR11607-5G
3-Chlor-6-(3-methoxypropylamino)pyridazin
3-Chlor-6-(3-methoxypropylamino)pyridazin
Formel: C₈H₁₂ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD08363096
CAS-Nummer: 898656-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR