4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester

APOSOR11595-1GEA 0 EUR
APOSOR11595-1G
4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester
4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester
Formel: C₁₁H₁₄BNO₃
Molecular Weight: 219,0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801045
CAS-Nummer: 957120-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR