5-Brom-4-cyclopentylpyrimidin

Lieferant: Apollo Scientific

APOSOR11578-5GEA 313 EUR
APOSOR11578-5G APOSOR11578-1G
5-Brom-4-cyclopentylpyrimidin
5-Bromo-4-cyclopentylpyrimidine
Formel: C₉H₁₁BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800933
CAS-Nummer: 951884-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR