Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid

APOSOR11575-5GEA 0 EUR
APOSOR11575-5G APOSOR11575-1G
Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid
Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid
Formel: C₁₀H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800986
CAS-Nummer: 957120-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR