5-Brom-4-propylpyrimidin

Lieferant: Apollo Scientific
Achtung

APOSOR11573-100MGEA 32.6 EUR
APOSOR11573-100MG APOSOR11573-1G
5-Brom-4-propylpyrimidin
5-Bromo-4-propylpyrimidine
Formel: C₇H₉BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800929
CAS-Nummer: 951884-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR