3-Brom-N-methylbenzamid 98%

Lieferant: Apollo Scientific
Achtung

APOSOR11565-250MGEA 15.1 EUR
APOSOR11565-250MG APOSOR11565-1G APOSOR11565-5G
3-Brom-N-methylbenzamid 98%
3-Brom-N-methylbenzamid
Formel: C₈H₈BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01060879
CAS-Nummer: 49834-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR