3-Brom-N-tert-butylbenzamid 97%
|
Achtung
|
APOSOR11560-1GEA
0
EUR
APOSOR11560-1G
APOSOR11560-5G
3-Brom-N-tert-butylbenzamid 97%
3-Brom-N-tert-butylbenzamid
Formel:
C₁₁H₁₄BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00751498 CAS-Nummer: 42498-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...