2-Brom-N-(2,3-dimethylphenyl)benzolsulfonamid 98%
Lieferant: Apollo Scientific
APOSOR11528-1GEA
102
EUR
APOSOR11528-1G
APOSOR11528-5G
2-Brom-N-(2,3-dimethylphenyl)benzolsulfonamid 98%
2-Brom-N-(2,3-dimethylphenyl)benzolsulfonamid
Formel:
C₁₄H₁₄BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD05617811 CAS-Nummer: 941294-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...