3-Brom-N-cyclohexyl-5-nitrobenzamid 98%

Lieferant: Apollo Scientific

APOSOR11497-5GEA 205 EUR
APOSOR11497-5G APOSOR11497-1G
3-Brom-N-cyclohexyl-5-nitrobenzamid 98%
3-Brom-N-cyclohexyl-5-nitrobenzamid
Formel: C₁₃H₁₅BrN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09475857
CAS-Nummer: 941294-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR