3-Brom-N-cyclopentyl-5-nitrobenzamid 97%
APOSOR11491-5GEA
0
EUR
APOSOR11491-5G
APOSOR11491-1G
3-Brom-N-cyclopentyl-5-nitrobenzamid 97%
3-Brom-N-cyclopentyl-5-nitrobenzamid
Formel:
C₁₂H₁₃BrN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475860 CAS-Nummer: 941294-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...