2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid 98%
APOSOR11470-1GEA
0
EUR
APOSOR11470-1G
APOSOR11470-5G
2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid 98%
2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid
Formel:
C₁₄H₁₄BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD05617812 CAS-Nummer: 941294-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...