Methyl-3-(3-boronobenzamido)propionat 97%

Lieferant: Apollo Scientific

APOSOR11433-1GEA 157 EUR
APOSOR11433-1G
Methyl-3-(3-boronobenzamido)propionat 97%
Methyl-3-(3-boronobenzamido)propionat
Formel: C₁₁H₁₄BNO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD09475827
CAS-Nummer: 957034-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR