3-Chlor-6-(1-imidazolyl)pyridazin 96%
|
Achtung
|
APOSOR11420-1GEA
0
EUR
APOSOR11420-1G
APOSOR11420-5G
3-Chlor-6-(1-imidazolyl)pyridazin 96%
3-Chloro-6-(1-imidazolyl)pyridazine
Formel:
C₇H₅ClN₄ Schmelzpunkt: 182…187 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02323226 CAS-Nummer: 71037-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...