3,6-Di(imidazol-1-yl)pyridazin 98%
APOSOR11418-5GEA
0
EUR
APOSOR11418-5G
APOSOR11418-1G
3,6-Di(imidazol-1-yl)pyridazin 98%
3,6-Di(imidazol-1-yl)pyridazin
Formel:
C₁₀H₈N₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475834 CAS-Nummer: 177648-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...