4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

APOSOR11404-1GEA 176 EUR
APOSOR11404-1G APOSOR11404-5G
4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid 97%
4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid
Formel: C₉H₈BrN₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09475848
CAS-Nummer: 957034-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR