4-Brom-1-(2-nitrophenyl)-1H-pyrazol 97%

APOSOR11403-1GEA 0 EUR
APOSOR11403-1G APOSOR11403-5G
4-Brom-1-(2-nitrophenyl)-1H-pyrazol 97%
4-Bromo-1-(2-nitrophenyl)-1H-pyrazole
Formel: C₉H₆BrN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09475850
CAS-Nummer: 957034-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR