4-Brom-1-(3-chlorphenyl)-1H-pyrazol 97%

Lieferant: Apollo Scientific

Achtung

APOSOR11402-1GEA 109 EUR
APOSOR11402-1G APOSOR11402-5G
4-Brom-1-(3-chlorphenyl)-1H-pyrazol 97%
4-Bromo-1-(3-chlorophenyl)-1H-pyrazole
Formel: C₉H₆BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09475849
CAS-Nummer: 957034-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR