2-Methoxyisonicotinsäurehydrazid
Lieferant: Apollo Scientific
APOSOR110761-1GEA
0
EUR
APOSOR110761-1G
2-Methoxyisonicotinsäurehydrazid
2-Methoxyisonicotinsäurehydrazid
Formel:
C₇H₉N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01747192 CAS-Nummer: 19353-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...