4-Amino-1-ethyl-1H-pyrazol

Lieferant: Apollo Scientific

Achtung

APOSOR110643-1GEA 85.8 EUR
APOSOR110643-1G
4-Amino-1-ethyl-1H-pyrazol
4-Amino-1-ethyl-1H-pyrazole
Formel: C₅H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05861667
CAS-Nummer: 876343-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR