4-(Ethoxycarbonyl)phenylboronsäurepinakolester 95+%

Lieferant: Apollo Scientific

APOSOR10989-5GEA 115 EUR
APOSOR10989-5G
4-(Ethoxycarbonyl)phenylboronsäurepinakolester 95+%
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03453659
CAS-Nummer: 195062-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR