4-Methoxycarbonyl-2-nitrophenylboronsäure 97%

Lieferant: Apollo Scientific

APOSOR10533-10GEA 82.9 EUR
APOSOR10533-10G
4-Methoxycarbonyl-2-nitrophenylboronsäure 97%
4-Methoxycarbonyl-2-nitrophenylboronsäure
Formel: C₈H₈BNO₆
Molecular Weight: 224,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632202
CAS-Nummer: 85107-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR