4-(Isobutylcarbamoyl)phenylboronsäure 97%
|
Achtung
|
APOSOR10518-5GEA
0
EUR
APOSOR10518-5G
APOSOR10518-1G
4-(Isobutylcarbamoyl)phenylboronsäure 97%
4-(Isobutylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221,06 g/mol Schmelzpunkt: 206…216 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411955 CAS-Nummer: 850568-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...