4-Iod-1-methylimidazol-5-carbaldehyd

Lieferant: Apollo Scientific

APOSOR10513-250MGEA 394 EUR
APOSOR10513-250MG
4-Iod-1-methylimidazol-5-carbaldehyd
4-Iod-1-methylimidazol-5-carbaldehyd
Formel: C₅H₅IN₂O
Molecular Weight: 236,0105 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01632219
CAS-Nummer: 189014-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR