3-Carboxyphenylboronsäurepinakolester
|
Achtung
|
APOSOR10405-1GEA
0
EUR
APOSOR10405-1G
APOSOR10405-5G
APOSOR10405-25G
3-Carboxyphenylboronsäurepinakolester
3-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molecular Weight: 248,083 g/mol Schmelzpunkt: 206…210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411930 CAS-Nummer: 269409-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...