3-tert-Butyl-5-methylphenylboronsäure
|
Achtung
|
APOSOR10396-250MGEA
0
EUR
APOSOR10396-250MG
APOSOR10396-1G
3-tert-Butyl-5-methylphenylboronsäure
3-tert-Butyl-5-methylphenylboronsäure
Formel:
C₁₁H₁₇BO₂ Schmelzpunkt: 158…160 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411938 CAS-Nummer: 193905-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...