3-(Benzyloxy)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR10361-1GEA
14.7
EUR
APOSOR10361-1G
APOSOR10361-25G
APOSOR10361-5G
3-(Benzyloxy)phenylboronsäure
3-(Benzyloxy)phenylboronsäure
Formel:
C₁₃H₁₃BO₃ Molecular Weight: 228,06 Schmelzpunkt: 125…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093063 CAS-Nummer: 156682-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...