2-[(Trimethylsilyl)ethinyl]benzaldehyd
|
Achtung
|
APOSOR01511-5GEA
0
EUR
APOSOR01511-5G
APOSOR01511-25G
2-[(Trimethylsilyl)ethinyl]benzaldehyd
2-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel:
C₁₂H₁₄OSi Schmelzpunkt: 48…53 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039984 CAS-Nummer: 77123-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...