4-Brom-6-methylchinolin 98%
APOSOR015007-5GEA
0
EUR
APOSOR015007-5G
APOSOR015007-1G
4-Brom-6-methylchinolin 98%
4-Bromo-6-methylquinoline
Formel:
C₁₀H₈BrN Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505108 CAS-Nummer: 1070879-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...