Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')barium Hydrat 99,9%

Lieferant: Apollo Scientific

APOSIN6578-10GEA 306 EUR
APOSIN6578-10G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')barium Hydrat 99,9%
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')barium Hydrat
Formel: C₂₂H₃₈BaO₄·H₂O
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200255
CAS-Nummer: 17594-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR