Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) 99.99%

Lieferant: Apollo Scientific

APOSIN5732-10GEA 241 EUR
APOSIN5732-10G
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) 99.99%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV)
Formel: C₄₄H₇₆O₈Zr
Molecular Weight: 824,29 g/mol
Schmelzpunkt: 308 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00145380
CAS-Nummer: 18865-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR