Digadoliniumtrioxid 99.99%
|
Achtung
|
Synonyme:
Gadolinium(III)oxid
APOSIN1837-250GEA
0
EUR
APOSIN1837-250G
APOSIN1837-50G
Digadoliniumtrioxid 99.99%
Digadoliniumtrioxid
Formel:
Gd₂O₃ Molecular Weight: 362,5 g/mol Schmelzpunkt: 2330 °C Dichte: 7,407 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011026 CAS-Nummer: 12064-62-9 EINECS: 235-060-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...