Dysprosium(III)carbonat Tetrahydrat 99.99%

Lieferant: Apollo Scientific

APOSIN1636-25GEA 0 EUR
APOSIN1636-25G
Dysprosium(III)carbonat Tetrahydrat 99.99%
Dysprosium(III)carbonat Tetrahydrat
Formel: C₃Dy₂O₉·4H₂O
Molecular Weight: 577,088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150229
CAS-Nummer: 38245-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR