Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')blei(II) 99,9%
APOSIN0954-10GEA
227
EUR
APOSIN0954-10G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')blei(II) 99,9%
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')blei(II)
Formel:
C₂₂H₃₈O₄Pb Molecular Weight: 573,73 g/mol Schmelzpunkt: 123 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00070468 CAS-Nummer: 21319-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...