N,N-Dimethylformamid-[D1] (>99,00% D)

APOSDE795-5MLEA 219 EUR
APOSDE795-5ML
N,N-Dimethylformamid-[D1] (>99,00% D)
N,N-Dimethylformamid-[D1]
Formel: DCOON(CH₃)₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2914-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR