Biotin-PEG4-Azide

APOSBIPG1217-100MGEA 0 EUR
APOSBIPG1217-100MG
Biotin-PEG4-Azide
Biotin-PEG4-Azide
Formel: C₂₀H₃₆N₆O₆S
Molecular Weight: 488,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29495433
CAS-Nummer: 1309649-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR