Okadasäure Kaliumsalz ≥98%

Lieferant: Apollo Scientific
Gefahr

APOSBIM1578-50UGEA 0 EUR
APOSBIM1578-50UG APOSBIM1578-1MG APOSBIM1578-100UG
Okadasäure Kaliumsalz ≥98%
Okadaic acid potassium salt

Potent inhibitor of protein phosphatases, especially the PP-1 and PP-2 classes, in numerous cell types.

Formel: C₄₄H₆₇O₁₃·K
Molecular Weight: 843,11 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00214356
CAS-Nummer: 155751-72-7
UN: 3462
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR