Tris-Glycine 10X Running Buffer (TG)
Lieferant: Apollo
APOSBI2043-10PACKSEA 0 EUR
APOSBI2043-10PACKS APOSBI2043-5PACKS
Tris-Glycine 10X Running Buffer (TG)
Puffer Elektrophorese-Puffer Laufpuffer
Electrophoresis running buffer.
Pre-weighed sachet containing sufficient buffer salts to create 1 litre of solution containing 250mM TRIS and 1.92M Glycine.
Bitte beachten Store at ambient temperatures.