6-Bromo-2-(chloromethyl)quinazoline
Lieferant: Apollo Scientific
APOSOR471371-1GEA
1010
EUR
APOSOR471371-1G
6-Bromo-2-(chloromethyl)quinazoline
6-Bromo-2-(chloromethyl)quinazoline
Formel:
C₉H₆BrClN₂ Molecular Weight: 257,514 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977356 CAS-Nummer: 1936538-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...