2-(2-Chloroethyl)-6-methoxyquinazoline

Lieferant: Apollo Scientific

APOSOR471367-1GEA 855 EUR
APOSOR471367-1G
2-(2-Chloroethyl)-6-methoxyquinazoline
2-(2-Chloroethyl)-6-methoxyquinazoline
Formel: C₁₁H₁₁ClN₂O
Molecular Weight: 222,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977360
CAS-Nummer: 2006277-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR